Assay Details
Target Gene Details
Entrez Gene ID: | 403178 |
Gene Name: | phosphatidylinositol-specific phospholipase C, X domain containing 1 |
Gene Aliases: |
A330045H12Rik, Gm576 |
Location: |
Chr.5:110099225-110105953 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plcxd1 | NM_001281812.1 | 3 | 219 | NP_001268741.1 |
NM_207279.2 | 4 | 386 | NP_997162.2 | |
AK039459.1 | 2 | 126 | BAC30356.1 | |
AK149435.1 | 3 | 220 | ||
BC039627.1 | 4 | 386 | AAH39627.1 |
