Assay Details
Target Gene Details
Entrez Gene ID: | 50791 |
Gene Name: | membrane associated guanylate kinase, WW and PDZ domain containing 2 |
Gene Aliases: |
AIP-1, Acvri1, Acvrinp1, Acvrip1, Magi-2, S-SCAM, mKIAA0705 |
Location: |
Chr.5:19223283-20704792 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Magi2 | NM_001170745.1 | NP_001164216.1 | ||
| NM_001170746.1 | NP_001164217.1 | |||
| NM_015823.3 | NP_056638.1 | |||
| AK039336.1 | BAC30321.1 | |||
| AK122349.1 | BAC65631.3 | |||
| AK147530.1 | BAE27976.1 | |||
| AY512951.1 | 2 | 3676 | AAR87821.1 | |
| BC059005.1 | AAH59005.1 |
More Information
Set Membership: |
|

Genomic Map