Assay Details
Target Gene Details
Entrez Gene ID: | 18861 |
Gene Name: | postmeiotic segregation increased 2 (S. cerevisiae) |
Gene Aliases: |
AW555130, Pmsl2 |
Location: |
Chr.5:143909974-143985719 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pms2 | NM_008886.2 | NP_032912.2 | ||
| AK017847.1 | ||||
| AK034866.1 | ||||
| AK083049.1 | ||||
| AK146365.1 | BAE27115.1 | |||
| BC139774.1 | ||||
| BC141287.1 |
More Information
Set Membership: |
|

Genomic Map