Assay Details
Target Gene Details
Entrez Gene ID: | 24116 |
Gene Name: | negative elongation factor complex member A, Whsc2 |
Gene Aliases: |
Whsc2, Whsc2h |
Location: |
Chr.5:33898179-33936258 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nelfa | NM_011914.2 | 7 | 907 | NP_036044.1 |
| AF101435.1 | 7 | 865 | AAC72983.1 | |
| AK019191.1 | 7 | 973 | ||
| AK078760.1 | 7 | 907 | BAC37379.1 | |
| AK088384.1 | 7 | 923 | BAC40319.1 | |
| AK146185.1 | 7 | 959 | BAE26961.1 | |
| AK180606.1 | 3 | 174 | ||
| BC038003.1 | 7 | 908 | AAH38003.1 |

Genomic Map