Assay Details
Target Gene Details
Entrez Gene ID: | 108151 |
Gene Name: | sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3D |
Gene Aliases: |
4631426B19Rik |
Location: |
Chr.5:12383166-12588943 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sema3d | NM_028882.4 | 9 | 1456 | NP_083158.3 |
| AK028900.1 | 9 | 1238 | BAC26185.1 | |
| AK031704.1 | 9 | 1068 | BAC27522.1 | |
| AK051165.1 | 9 | 1117 | BAC34542.1 | |
| AK052671.1 | 9 | 1457 | BAC35092.1 | |
| AK153839.1 | 9 | 1117 | ||
| BC138131.1 | 9 | 999 |
More Information
Set Membership: |
|

Genomic Map