Assay Details
Target Gene Details
Entrez Gene ID: | 214642 |
Gene Name: | cadherin-like and PC-esterase domain containing 1 |
Gene Aliases: |
6720481P07, A430107O13Rik, AI552584 |
Location: |
Chr.6:21985705-22256407 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cped1 | NM_001081351.1 | NP_001074820.1 | ||
| AK032959.1 | BAC28099.1 | |||
| BC151018.1 |

Genomic Map