Assay Details
Target Gene Details
Entrez Gene ID: | 21899 |
Gene Name: | toll-like receptor 6 |
Gene Aliases: |
- |
Location: |
Chr.5:64952030-64971173 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tlr6 | NM_011604.3 | NP_035734.3 | ||
| AK154253.1 | ||||
| BC055366.1 | AAH55366.1 |

Genomic Map