Assay Details
Target Gene Details
Entrez Gene ID: | 83704 |
Gene Name: | solute carrier family 12 (potassium/chloride transporters), member 9 |
Gene Aliases: |
CIP1 |
Location: |
Chr.5:137314558-137335688 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc12a9 | NM_031406.3 | 2 | 209 | NP_113583.2 |
| AF314957.1 | 1 | 75 | AAL26866.1 | |
| AK050658.1 | 2 | 224 | ||
| AK154118.1 | 2 | 167 | ||
| BC046982.1 | 2 | 129 | AAH46982.1 |
More Information
Set Membership: |
|

Genomic Map