Assay Details
Target Gene Details
Entrez Gene ID: | 22171 |
Gene Name: | thymidylate synthase |
Gene Aliases: |
Ts |
Location: |
Chr.5:30058200-30073625 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tyms | NM_021288.4 | NP_067263.1 | ||
| NR_033402.1 | ||||
| AK011435.1 | BAB27620.1 | |||
| AK035663.1 | BAC29144.1 | |||
| AK088134.1 | BAC40165.1 | |||
| AK088606.1 | BAC40450.1 | |||
| AK164981.1 | ||||
| AK207243.1 | ||||
| BC020139.1 | AAH20139.1 | |||
| M13019.1 | AAA40439.1 | |||
| X14489.1 | 2 | 1063 | CAA32651.1 |
More Information
Set Membership: |
|

Genomic Map