Assay Details
Target Gene Details
Entrez Gene ID: | 67956 |
Gene Name: | SET domain containing (lysine methyltransferase) 8 |
Gene Aliases: |
2410195B05Rik, AA617402, AW536475, PR-SET7 |
Location: |
Chr.5:124439930-124462308 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Setd8 | NM_030241.2 | NP_084517.2 | ||
| AK010593.1 | BAC25304.1 | |||
| AK010835.1 | ||||
| AK030904.1 | BAC27178.1 | |||
| AK088097.1 | ||||
| BB619898.1 | ||||
| BC108333.1 | AAI08334.1 |
More Information
Set Membership: |
|

Genomic Map