Assay Details
Target Gene Details
Entrez Gene ID: | 108014 |
Gene Name: | serine/arginine-rich splicing factor 9 |
Gene Aliases: |
25kDa, 2610029M16Rik, SRp30c, Sfrs9 |
Location: |
Chr.5:115327161-115333084 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srsf9 | NM_025573.3 | NP_079849.1 | ||
| NR_036616.1 | ||||
| AK011621.1 | BAB27740.1 | |||
| AK020084.1 | BAB31986.1 | |||
| AK159190.1 | BAE34886.1 | |||
| BC012217.1 | AAH12217.1 | |||
| CJ162734.1 | ||||
| CV558546.1 |
More Information
Set Membership: |
|

Genomic Map