Assay Details
Target Gene Details
Entrez Gene ID: | 76571 |
Gene Name: | serine/threonine/tyrosine interacting-like 1 |
Gene Aliases: |
1700011C14Rik, Dusp24, Mkstyx |
Location: |
Chr.5:135747220-135778410 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Styxl1 | NM_001289554.1 | 7 | 803 | NP_001276483.1 |
NM_001289555.1 | 7 | 800 | NP_001276484.1 | |
NM_001289556.1 | 6 | 644 | NP_001276485.1 | |
NM_001289557.1 | 5 | 495 | NP_001276486.1 | |
NM_029659.4 | 7 | 810 | NP_083935.3 | |
AK005599.1 | 7 | 801 | BAB24144.1 | |
AK005858.1 | 6 | 551 | BAB24279.1 | |
AK161310.1 | 7 | 811 | BAE36314.1 | |
BC045148.1 | 7 | 740 | AAH45148.1 | |
BF471066.1 | 5 | 404 |
