Assay Details
Target Gene Details
Entrez Gene ID: | 67690 |
Gene Name: | protease, serine 37 |
Gene Aliases: |
1700016G05Rik, Tryx2 |
Location: |
Chr.6:40514820-40519542 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss37 | NM_026317.2 | 2 | 182 | NP_080593.1 |
| AK006028.1 | 2 | 186 | BAB24373.1 | |
| AK016088.1 | 2 | 187 | BAB30109.1 | |
| AK029780.1 | 2 | 159 | BAC26613.1 | |
| BC125467.1 | ||||
| BC125493.1 |
More Information
Set Membership: |
|

Genomic Map