Assay Details
Target Gene Details
Entrez Gene ID: | 109552 |
Gene Name: | sorcin |
Gene Aliases: |
2210417O06Rik, 2900070H08Rik, Sor |
Location: |
Chr.5:8046078-8069314 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sri | NM_025618.3 | NP_079894.2 | ||
AK008404.1 | BAB25652.1 | |||
AK008783.1 | BAB25891.1 | |||
AK008970.1 | BAB25997.1 | |||
BY708689.1 |
