Assay Details
Target Gene Details
Entrez Gene ID: | 109552 |
Gene Name: | sorcin |
Gene Aliases: |
2210417O06Rik, 2900070H08Rik, Sor |
Location: |
Chr.5:8046078-8069314 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sri | NM_001080974.2 | NP_001074443.1 | ||
NM_025618.3 | NP_079894.2 | |||
AK008404.1 | BAB25652.1 | |||
AK008783.1 | BAB25891.1 | |||
AK008970.1 | BAB25997.1 | |||
AK013762.1 | 1 | 1141 | ||
AK134747.1 | BAE22266.1 | |||
AK146071.1 | BAE26876.1 | |||
AK146911.1 | BAE27523.1 | |||
AK168572.1 | BAE40442.1 | |||
AK168792.1 | BAE40625.1 | |||
AK168900.1 | BAE40716.1 | |||
BC065790.1 | AAH65790.1 | |||
BY708689.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
