Assay Details
Target Gene Details
Entrez Gene ID: | 20817 |
Gene Name: | serine/arginine-rich protein specific kinase 2 |
Gene Aliases: |
AW226533, AW492537, AW547358, Wbp6, mSRPK2 |
Location: |
Chr.5:23503356-23684641 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srpk2 | NM_009274.2 | NP_033300.2 | ||
| AB006036.1 | BAA24055.1 | |||
| AK014004.1 | ||||
| AK136861.1 | ||||
| AK161384.1 | BAE36363.1 | |||
| AK169580.1 | BAE41239.1 | |||
| BB860938.1 | ||||
| BC020178.1 | AAH20178.1 | |||
| BC062941.1 | AAH62941.1 | |||
| U92456.1 | AAC53193.1 |
More Information
Set Membership: |
|

Genomic Map