Assay Details
Target Gene Details
Entrez Gene ID: | 100763 |
Gene Name: | ubiquitin protein ligase E3C |
Gene Aliases: |
AI853514, mKIAA0010 |
Location: |
Chr.5:29569242-29676077 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube3c | NM_133907.3 | 9 | 1336 | NP_598668.1 |
| AK029973.1 | 9 | 1146 | BAC26709.1 | |
| AK045211.1 | 9 | 1134 | ||
| AK046056.1 | 9 | 1337 | BAC32585.1 | |
| AK122187.1 | 9 | 1271 | BAC65469.1 | |
| BC120731.1 | 9 | 1083 | ||
| BC137626.1 | 9 | 1083 |
More Information
Set Membership: |
|

Genomic Map