Assay Details
Target Gene Details
Entrez Gene ID: | 20817 |
Gene Name: | serine/arginine-rich protein specific kinase 2 |
Gene Aliases: |
AW226533, AW492537, AW547358, Wbp6, mSRPK2 |
Location: |
Chr.5:23503356-23684641 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srpk2 | NM_009274.2 | 9 | 1065 | NP_033300.2 |
| AB006036.1 | 9 | 858 | BAA24055.1 | |
| AK136861.1 | 9 | 1031 | ||
| AK196829.1 | 3 | 181 | ||
| BC020178.1 | 9 | 962 | AAH20178.1 | |
| BC062941.1 | 9 | 984 | AAH62941.1 |
More Information
Set Membership: |
|

Genomic Map