Assay Details
Target Gene Details
Entrez Gene ID: | 14311 |
Gene Name: | cell death-inducing DFFA-like effector c |
Gene Aliases: |
CIDE-3, Fsp27 |
Location: |
Chr.6:113424634-113435760 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cidec | NM_001301295.1 | NP_001288224.1 | ||
| NM_178373.3 | NP_848460.1 | |||
| AK080133.1 | BAC37830.1 | |||
| BC099676.1 | AAH99676.1 | |||
| M61737.1 |

Genomic Map