Assay Details
Target Gene Details
Entrez Gene ID: | 11628 |
Gene Name: | activation-induced cytidine deaminase |
Gene Aliases: |
Aid, Arp2 |
Location: |
Chr.6:122553809-122564180 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aicda | NM_009645.2 | NP_033775.1 | ||
| AF132979.1 | AAD41793.1 | |||
| CJ161825.1 |
More Information
Set Membership: |
|

Genomic Map