Assay Details
Target Gene Details
Entrez Gene ID: | 11677 |
Gene Name: | aldo-keto reductase family 1, member B3 (aldose reductase) |
Gene Aliases: |
ALR2, AR, Ahr-1, Ahr1, Akr1b1, Aldor1, Aldr1 |
Location: |
Chr.6:34303930-34317489 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr1b3 | NM_009658.3 | NP_033788.3 | ||
| AK149861.1 | BAE29130.1 | |||
| AK153949.1 | BAE32272.1 | |||
| AK170663.1 | BAE41944.1 | |||
| BC021655.1 | AAH21655.1 | |||
| BY749711.1 |
More Information
Set Membership: |
|

Genomic Map