Assay Details
Target Gene Details
Entrez Gene ID: | 14056 |
Gene Name: | enhancer of zeste homolog 2 (Drosophila) |
Gene Aliases: |
Enx-1, Enx1h, KMT6, mKIAA4065 |
Location: |
Chr.6:47530274-47595340 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ezh2 | NM_001146689.1 | NP_001140161.1 | ||
| NM_007971.2 | NP_031997.2 | |||
| AK088575.1 | BAC40432.2 | |||
| AK153835.1 | BAE32205.1 | |||
| AK157853.1 | BAE34232.1 | |||
| AK220174.1 | BAD90359.1 | |||
| BC003772.1 | AAH03772.1 | |||
| BC016391.1 | AAH16391.1 | |||
| BC079538.1 | AAH79538.1 | |||
| U52951.1 | AAC52655.1 |
More Information
Set Membership: |
|

Genomic Map