Assay Details
Target Gene Details
Entrez Gene ID: | 11993 |
Gene Name: | ancient ubiquitous protein 1 |
Gene Aliases: |
AA589454 |
Location: |
Chr.6:83054521-83057682 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aup1 | NM_001301649.1 | 3 | 498 | NP_001288578.1 |
| NM_007517.4 | 4 | 649 | NP_031543.2 | |
| NR_125891.1 | 4 | 649 | ||
| AK144497.1 | 1 | 826 | ||
| AK154707.1 | 4 | 518 | BAE32776.1 | |
| AK158035.1 | 4 | 440 | BAE34327.1 | |
| AK167335.1 | 4 | 439 | ||
| AK171011.1 | 4 | 452 | BAE42182.1 | |
| AK171184.1 | 4 | 444 | ||
| AK171202.1 | 4 | 458 | ||
| AK171249.1 | 4 | 438 | ||
| AK171541.1 | 4 | 439 | BAE42515.1 | |
| AK171621.1 | 4 | 431 | ||
| AK180069.1 | ||||
| AK195286.1 | ||||
| AK209976.1 | ||||
| BC016485.1 | 4 | 442 | AAH16485.1 | |
| U41736.1 | 4 | 447 | AAC52839.1 |
More Information
Set Membership: |
|

Genomic Map