Assay Details
Target Gene Details
Entrez Gene ID: | 232413 |
Gene Name: | C-type lectin domain family 12, member a |
Gene Aliases: |
CLL-1, D230024O04, KLRL1, Micl |
Location: |
Chr.6:129349779-129365303 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clec12a | NM_177686.4 | NP_808354.1 | ||
| AK084335.1 | BAC39163.1 | |||
| BC094904.1 | AAH94904.1 |

Genomic Map