Assay Details
Target Gene Details
Entrez Gene ID: | 67755 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 47 |
Gene Aliases: |
4930588A18Rik, C77285 |
Location: |
Chr.6:135011612-135023776 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx47 | NM_026360.3 | 3 | 343 | NP_080636.2 |
AK010310.1 | 3 | 336 | BAB26843.2 | |
AK076982.1 | 3 | 328 | BAC36547.1 | |
BC095944.1 | 3 | 320 | AAH95944.1 | |
BC132242.1 | 3 | 335 | ||
BC132244.1 | 3 | 335 | ||
BY123864.1 |
