Assay Details
Target Gene Details
Entrez Gene ID: | 232406 |
Gene Name: | cDNA sequence BC035044 |
Gene Aliases: |
- |
Location: |
Chr.6:128848044-128891124 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BC035044 | NM_001254946.1 | NP_001241875.1 | ||
| NM_001254947.1 | NP_001241876.1 | |||
| AK088258.1 |
Target Gene Details
Entrez Gene ID: | 93675 |
Gene Name: | C-type lectin domain family 2, member i |
Gene Aliases: |
Clr-g, Clrg, Dcl1, LCL-1, OCILrP2 |
Location: |
Chr.6:128887573-128898167 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clec2i | NM_001289706.1 | NP_001276635.1 | ||
| NM_001289707.1 | NP_001276636.1 | |||
| NM_001289708.1 | NP_001276637.1 | |||
| NM_020257.2 | NP_064653.1 | |||
| AF121352.1 | AAD22055.1 | |||
| AF350411.1 | AAK70359.1 | |||
| AK041724.1 | ||||
| AY137344.1 | AAN15953.1 | |||
| AY137345.1 | AAN15954.1 | |||
| AY256574.1 | AAP32744.1 | |||
| AY256575.1 | AAP32745.1 | |||
| AY256576.1 | AAP32746.1 | |||
| BC139192.1 | ||||
| BC139193.1 | ||||
| BC145574.1 |
More Information
Set Membership: |
|

Genomic Map