Assay Details
Target Gene Details
Entrez Gene ID: | 12576 |
Gene Name: | cyclin-dependent kinase inhibitor 1B |
Gene Aliases: |
AA408329, AI843786, Kip1, p27, p27Kip1 |
Location: |
Chr.6:134920401-134925525 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdkn1b | NM_009875.4 | NP_034005.2 | ||
| AK046676.1 | 2 | 836 | BAC32833.1 | |
| AK047669.1 | 2 | 1102 | BAC33119.1 | |
| AK050240.1 | 2 | 1088 | BAC34141.1 | |
| AK158411.1 | 2 | 1081 | BAE34494.1 | |
| AK162146.1 | ||||
| BC014296.1 | AAH14296.1 | |||
| CT010382.1 | CAJ18589.1 | |||
| U09968.1 | AAA20235.1 | |||
| U10440.1 | AAA21149.1 |
More Information
Set Membership: |
|

Genomic Map