Assay Details
Target Gene Details
Entrez Gene ID: | 69810 |
Gene Name: | C-type lectin domain family 4, member b1 |
Gene Aliases: |
1810046I24Rik, Aplra2, Clec4b, DCAR, DCARbeta, Dcar2 |
Location: |
Chr.6:123049962-123071555 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clec4b1 | NM_001190310.1 | NP_001177239.1 | ||
| NM_027218.3 | NP_081494.1 | |||
| AA914236.1 | ||||
| AK007794.1 | BAB25260.1 | |||
| AY230259.1 | AAP58453.1 | |||
| AY230260.1 | AAP58454.1 | |||
| AY365134.1 | AAR13070.1 |
More Information
Set Membership: |
|

Genomic Map