Assay Details
Target Gene Details
Entrez Gene ID: | 12638 |
Gene Name: | cystic fibrosis transmembrane conductance regulator |
Gene Aliases: |
AW495489, Abcc7 |
Location: |
Chr.6:18170262-18322774 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cftr | NM_021050.2 | NP_066388.1 | ||
| M60493.1 | AAA18903.1 | |||
| M69298.1 | AAA37417.1 |
More Information
Set Membership: |
|

Genomic Map