Assay Details
Target Gene Details
Entrez Gene ID: | 320127 |
Gene Name: | diacylglycerol kinase, iota |
Gene Aliases: |
C130010K08Rik |
Location: |
Chr.6:36837381-37300060 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dgki | NM_001081206.1 | NP_001074675.1 | ||
| AK081360.1 | ||||
| AK086550.1 |
More Information
Set Membership: |
|

Genomic Map