Assay Details
Target Gene Details
Entrez Gene ID: | 56620 |
Gene Name: | C-type lectin domain family 4, member n |
Gene Aliases: |
Clec6a, Clecsf10, Nkcl |
Location: |
Chr.6:123229843-123247024 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clec4n | NM_001190320.1 | NP_001177249.1 | ||
| NM_001190321.1 | NP_001177250.1 | |||
| NM_020001.2 | NP_064385.1 | |||
| AF240357.1 | AAF67177.1 | |||
| AF240358.1 | AAF67178.1 | |||
| AF240359.1 | AAF67179.1 | |||
| AK089166.1 | ||||
| AK131697.1 | BAE20766.1 | |||
| AK154673.1 | BAE32756.1 | |||
| AK171046.1 | ||||
| BC023008.1 | AAH23008.1 |
More Information
Set Membership: |
|

Genomic Map