Assay Details
Target Gene Details
Entrez Gene ID: | 66075 |
Gene Name: | coiled-coil-helix-coiled-coil-helix domain containing 3 |
Gene Aliases: |
0610041L09Rik, 1700039J09Rik, AW558177 |
Location: |
Chr.6:32792217-33060276 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chchd3 | NM_025336.1 | NP_079612.1 | ||
| AK002263.1 | BAB21974.1 | |||
| AK002889.1 | ||||
| AK003519.1 | BAB22832.1 | |||
| AK006643.1 | BAB24685.1 | |||
| AK050063.1 | 1 | 225 | ||
| AK145337.1 | BAE26374.1 | |||
| BC021941.1 | AAH21941.1 |

Genomic Map