Assay Details
Target Gene Details
Entrez Gene ID: | 73288 |
Gene Name: | coiled-coil domain containing 132 |
Gene Aliases: |
1700034M03Rik, 8430415E05Rik, BLM, C87205, mKIAA1861 |
Location: |
Chr.6:3498393-3603531 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc132 | NM_001167750.1 | NP_001161222.1 | ||
NM_001167751.1 | NP_001161223.1 | |||
NM_024260.5 | NP_077222.4 | |||
AK018415.1 | BAB31200.1 | |||
AK042031.1 | BAC31137.1 | |||
AK075898.1 | BAC36038.1 | |||
AK129458.1 | BAC98268.1 | |||
AY254697.1 | AAO84056.1 | |||
BC036294.1 | AAH36294.2 | |||
BC043092.1 | AAH43092.1 |
