Assay Details
Target Gene Details
Entrez Gene ID: | 56150 |
Gene Name: | MAD2 mitotic arrest deficient-like 1 |
Gene Aliases: |
AA673185, MAD2 |
Location: |
Chr.6:66535249-66541109 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mad2l1 | NM_019499.4 | NP_062372.2 | ||
| AK008638.1 | BAB25799.1 | |||
| AK010401.1 | ||||
| AK012590.1 | BAB28338.1 | |||
| AK082934.1 | BAC38700.1 | |||
| AK160286.1 | BAE35726.1 | |||
| AK166065.1 | ||||
| BC089012.1 | AAH89012.1 | |||
| U83902.1 | AAD09238.1 |

Genomic Map