Assay Details
Target Gene Details
Entrez Gene ID: | 101202 |
Gene Name: | HEPACAM family member 2 |
Gene Aliases: |
AI987662 |
Location: |
Chr.6:3457089-3498189 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hepacam2 | NM_178899.5 | NP_849230.1 | ||
| AK033728.1 | BAC28450.1 | |||
| AK050501.1 | BAC34294.1 | |||
| BC096374.1 | AAH96374.1 |
More Information
Set Membership: |
|

Genomic Map