Assay Details
Target Gene Details
Entrez Gene ID: | 93695 |
Gene Name: | glycoprotein (transmembrane) nmb |
Gene Aliases: |
DC-HIL, Dchil, ipd |
Location: |
Chr.6:49036518-49058182 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpnmb | NM_053110.4 | 5 | 822 | NP_444340.3 |
| AF322054.1 | 5 | 660 | AAK14240.1 | |
| AJ251685.1 | 5 | 707 | CAB65272.1 | |
| AK044764.1 | 5 | 716 | BAC32074.1 | |
| AK076347.1 | 5 | 691 | BAC36306.1 | |
| AK149700.1 | 5 | 691 | BAE29036.1 | |
| AK149740.1 | 5 | 729 | ||
| AK150739.1 | 5 | 691 | BAE29813.1 | |
| AK154617.1 | 5 | 726 | BAE32716.1 | |
| AK159165.1 | 5 | 721 | BAE34867.1 | |
| AK159270.1 | 5 | 729 | BAE34949.1 | |
| AK159747.1 | 5 | 691 | BAE35339.1 | |
| AK159908.1 | 5 | 697 | ||
| AK170506.1 | 5 | 721 | BAE41844.1 | |
| AK171347.1 | 5 | 697 | BAE42404.1 | |
| AK171441.1 | 5 | 691 | BAE42454.1 | |
| AK171465.1 | 5 | 691 | BAE42471.1 | |
| AK171474.1 | 5 | 691 | BAE42479.1 | |
| AK171517.1 | 5 | 809 | BAE42501.1 | |
| AK171608.1 | 5 | 691 | BAE42560.1 | |
| AK171618.1 | 5 | 691 | BAE42567.1 | |
| AK179971.1 | 2 | 99 | ||
| AK192937.1 | ||||
| AK193247.1 | ||||
| AK193314.1 | ||||
| AK193818.1 | ||||
| AK195422.1 | ||||
| AK196346.1 | ||||
| AK196951.1 | ||||
| AK201982.1 | ||||
| AK204190.1 | ||||
| AK214279.1 | ||||
| AK215880.1 | ||||
| BC026375.1 | 5 | 696 | AAH26375.1 |
More Information
Set Membership: |
|

Genomic Map