Assay Details
Target Gene Details
Entrez Gene ID: | 140488 |
Gene Name: | insulin-like growth factor 2 mRNA binding protein 3 |
Gene Aliases: |
2610101N11Rik, AA522010, AL022933, AU045931, IMP-3, IMP3, Koc13, Neilsen, mimp3 |
Location: |
Chr.6:49085218-49215009 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igf2bp3 | NM_023670.3 | NP_076159.3 | ||
| AB046173.1 | BAB19755.1 | |||
| AK011689.1 | BAB27779.1 | |||
| AK011797.1 | ||||
| AK030096.1 | ||||
| AK088465.1 | BAC40370.1 | |||
| AK147140.1 | BAE27710.1 | |||
| BC045138.1 | AAH45138.1 | |||
| BC049082.1 | AAH49082.1 |
More Information
Set Membership: |
|

Genomic Map