Assay Details
Target Gene Details
Entrez Gene ID: | 66441 |
Gene Name: | mago-nashi homolog B (Drosophila) |
Gene Aliases: |
2010012C16Rik, Mago, Magoh, Magoh-rs1 |
Location: |
Chr.6:131284388-131293244 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Magohb | NM_025564.2 | 1 | 127 | NP_079840.2 |
AK006303.1 | 1 | 117 | BAB24516.2 | |
AK007769.1 | 1 | 113 | BAB25244.1 | |
AK008200.1 | 1 | 129 | BAB25528.1 | |
AK013310.1 | 1 | 112 | BAB28784.1 | |
BC027505.1 | 1 | 100 | AAH27505.1 | |
KF051005.1 |
