Assay Details
Target Gene Details
Entrez Gene ID: | 338523 |
Gene Name: | lysine (K)-specific demethylase 7A |
Gene Aliases: |
A630082K20Rik, BB041802, ENSMUSG00000073143, Jhdm1d, mKIAA1718 |
Location: |
Chr.6:39136620-39206773 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kdm7a | NM_001033430.4 | NP_001028602.2 | ||
AK042327.1 | BAC31226.2 | |||
AK042834.1 | BAC31377.1 | |||
AK136209.1 | BAE22876.1 | |||
AK136238.1 | BAE22890.1 | |||
BC007161.1 | AAH07161.1 | |||
BC145848.1 |
