Assay Details
Target Gene Details
Entrez Gene ID: | 17193 |
Gene Name: | methyl-CpG binding domain protein 4 |
Gene Aliases: |
Med1 |
Location: |
Chr.6:115840697-115853438 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mbd4 | NM_010774.2 | NP_034904.2 | ||
| AF072249.1 | AAC68878.1 | |||
| AK147782.1 | BAE28134.1 | |||
| AK161005.1 | BAE36144.1 | |||
| BC024812.1 | AAH24812.1 |
More Information
Set Membership: |
|

Genomic Map