Assay Details
Target Gene Details
Entrez Gene ID: | 17992 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4 |
Gene Aliases: |
MLRQ |
Location: |
Chr.6:11900373-11907446 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufa4 | NM_010886.2 | NP_035016.1 | ||
| AK005084.1 | ||||
| AK008357.1 | BAB25627.1 | |||
| AK008506.1 | BAB25706.1 | |||
| AK076036.1 | BAC36137.1 | |||
| AK154084.1 | ||||
| BC011114.1 | AAH11114.1 |
More Information
Set Membership: |
|

Genomic Map