Assay Details
Target Gene Details
Entrez Gene ID: | 114142 |
Gene Name: | forkhead box P2 |
Gene Aliases: |
2810043D05Rik, AI449000, CAG-16, D0Kist7 |
Location: |
Chr.6:14901226-15441977 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Foxp2 | NM_053242.4 | 3 | 595 | NP_444472.2 |
| AK051668.1 | 3 | 557 | BAC34711.1 | |
| AK080549.1 | 3 | 466 | ||
| AK082361.1 | 3 | 267 | BAC38477.1 | |
| AY079003.1 | 3 | 224 | AAL85482.1 | |
| CN526766.1 |

Genomic Map