Assay Details
Target Gene Details
Entrez Gene ID: | 17059 |
Gene Name: | killer cell lectin-like receptor subfamily B member 1C |
Gene Aliases: |
AI462337, CD161, Ly-59, Ly55c, Ly59, NK-RP1, NK1.1, NKR-P1.9, NKRP1, NKRP140, Nk-1, Nk-1.2, Nk1, Nk1.2, Nkrp1c, ly-55c |
Location: |
Chr.6:128778485-128788641 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klrb1c | NM_001159904.1 | 1 | 133 | NP_001153376.1 |
| NM_008527.2 | 1 | 133 | NP_032553.2 | |
| AF354263.1 | AAK39103.1 | |||
| AK079743.1 | 1 | 135 | BAC37739.1 | |
| BC061168.1 | 1 | 87 | AAH61168.1 | |
| DQ237929.1 | ||||
| M77678.1 | 1 | 72 | AAA39824.1 | |
| S43141.1 | AAB22979.1 |
More Information
Set Membership: |
|

Genomic Map