Assay Details
Target Gene Details
Entrez Gene ID: | 232157 |
Gene Name: | MOB kinase activator 1A |
Gene Aliases: |
4022402H07Rik, MOB1, MOB4B, Mobk1b, Mobkl1b |
Location: |
Chr.6:83326033-83343696 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mob1a | NM_145571.2 | NP_663546.1 | ||
| AK028416.1 | 1 | 91 | BAC25938.1 | |
| AK028695.1 | BAC26070.1 | |||
| AK158228.1 | ||||
| AK163300.1 | 1 | 90 | ||
| AK167515.1 | BAE39589.1 | |||
| AK170691.1 | ||||
| AK172205.1 | BAE42880.1 | |||
| BC009149.1 | AAH09149.1 | |||
| BC033463.1 | AAH33463.1 |
More Information
Set Membership: |
|

Genomic Map