Assay Details
Target Gene Details
Entrez Gene ID: | 114142 |
Gene Name: | forkhead box P2 |
Gene Aliases: |
2810043D05Rik, AI449000, CAG-16, D0Kist7 |
Location: |
Chr.6:14901226-15441977 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Foxp2 | NM_001286607.1 | 7 | 1171 | NP_001273536.1 |
NM_053242.4 | 9 | 1571 | NP_444472.2 | |
NM_212435.1 | 7 | 1234 | NP_997600.1 | |
AF339106.1 | 6 | 912 | AAK69651.1 | |
AK051668.1 | 9 | 1554 | BAC34711.1 | |
AK082361.1 | 9 | 1243 | BAC38477.1 | |
AY079003.1 | 9 | 1200 | AAL85482.1 | |
BC058960.1 | 7 | 1168 | AAH58960.1 | |
BC062926.1 | 7 | 1234 | AAH62926.1 |
