Assay Details
Target Gene Details
Entrez Gene ID: | 56463 |
Gene Name: | staphylococcal nuclease and tudor domain containing 1 |
Gene Aliases: |
AL033314, Tudor-SN |
Location: |
Chr.6:28480315-28935162 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Snd1 | NM_019776.2 | 5 | 764 | NP_062750.2 |
AB021491.1 | 5 | 721 | BAA84944.1 | |
AK034914.1 | 4 | 633 | ||
AK088028.1 | 5 | 723 | BAC40105.1 | |
AK133837.1 | 5 | 770 | BAE21874.1 | |
AK159837.1 | 5 | 729 | BAE35415.1 | |
AK161591.1 | 5 | 718 | BAE36479.1 | |
AK162432.1 | 5 | 705 | BAE36913.1 | |
AK167576.1 | 5 | 754 | BAE39639.1 | |
AK178471.1 | 1 | 99 | ||
BC007126.1 | 5 | 711 | AAH07126.3 | |
BM944959.1 | 4 | 442 |
