Assay Details
Target Gene Details
Entrez Gene ID: | 56463 |
Gene Name: | staphylococcal nuclease and tudor domain containing 1 |
Gene Aliases: |
AL033314, Tudor-SN |
Location: |
Chr.6:28480315-28935162 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Snd1 | NM_019776.2 | 8 | 1102 | NP_062750.2 |
AB021491.1 | 8 | 1059 | BAA84944.1 | |
AK034914.1 | 7 | 971 | ||
AK088028.1 | 8 | 1061 | BAC40105.1 | |
AK133837.1 | 8 | 1108 | BAE21874.1 | |
AK159837.1 | 8 | 1067 | BAE35415.1 | |
AK161591.1 | 8 | 1056 | BAE36479.1 | |
AK162432.1 | 8 | 1043 | BAE36913.1 | |
AK167576.1 | 8 | 1092 | BAE39639.1 | |
AK183742.1 | ||||
AK185603.1 | ||||
AK190012.1 | ||||
AK196687.1 | ||||
AK200045.1 | ||||
AK216953.1 | ||||
AK220074.1 | ||||
BC007126.1 | 8 | 1049 | AAH07126.3 |
