Assay Details
Target Gene Details
Entrez Gene ID: | 21848 |
Gene Name: | tripartite motif-containing 24 |
Gene Aliases: |
A130082H20Rik, AI447469, D430004I05Rik, TIF1, TIF1-alpha, TIF1alpha, Tif1a |
Location: |
Chr.6:37869880-37968445 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim24 | NM_001272064.1 | 14 | 2369 | NP_001258993.1 |
| NM_001272076.1 | 14 | 2051 | NP_001259005.1 | |
| NM_145076.4 | 14 | 2471 | NP_659542.3 | |
| AB221637.1 | 14 | 2171 | ||
| AK133144.1 | 14 | 2051 | BAE21530.1 | |
| AK166538.1 | 10 | 1428 | BAE38840.1 | |
| BC025482.1 | 5 | 566 | AAH25482.1 | |
| BC056959.1 | 14 | 2449 | AAH56959.1 | |
| S78219.1 | 14 | 2369 | AAB34289.1 | |
| S78221.1 | 14 | 2471 | AAB34290.1 |
More Information
Set Membership: |
|

Genomic Map