Assay Details
Target Gene Details
Entrez Gene ID: | 66367 |
Gene Name: | RIKEN cDNA 2310022A10 gene |
Gene Aliases: |
- |
Location: |
Chr.7:27553260-27582099 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2310022A10Rik | NM_001122767.1 | NP_001116239.1 | ||
| NM_175107.5 | NP_780316.3 | |||
| AK009456.1 | BAC25259.1 | |||
| AK009941.1 | ||||
| AK077369.1 | BAC36772.1 | |||
| AK140814.1 | BAE24486.1 | |||
| BB853451.1 | ||||
| BC023369.1 | AAH23369.2 |

Genomic Map