Assay Details
Target Gene Details
Entrez Gene ID: | 641361 |
Gene Name: | phospholipase A2 inhibitor and LY6/PLAUR domain containing |
Gene Aliases: |
2310033E01Rik |
Location: |
Chr.7:24541654-24546313 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pinlyp | NM_001037143.2 | 3 | 241 | NP_001032220.1 |
| AK009589.1 | 3 | 246 | BAB26378.1 | |
| AK009604.1 | 3 | 243 | BAB26386.1 | |
| AK009778.1 | 3 | 241 | BAB26498.1 | |
| AK009924.1 | 3 | 243 | BAB26587.1 | |
| BC139039.1 | 3 | 307 | ||
| BC145686.1 | 3 | 307 |
More Information
Set Membership: |
|

Genomic Map